Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Antibody, Novus Biologicals™
SDP

Chicken Polyclonal Antibody has been used in 26 publications

$219.50 - $621.00

Specifications

Antigen mCherry
Dilution Western Blot 1:2000-1:5000, ELISA, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, IHC
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019394
SDP
View Documents
Novus Biologicals
NBP2251580.025ML
0.025 mL
Each for $219.50
Only null left
Add to Cart
 
NBP225158
SDP
View Documents
Novus Biologicals
NBP225158
0.1 mL
Each for $621.00
Only null left
Add to Cart
 
Description

Description

mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount.
Specifications

Specifications

mCherry
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Western Blot
Unconjugated
Chicken
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:2000-1:5000, ELISA, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, IHC
Polyclonal
Precipitation
RUO
IgY
Ammonium sulfate precipitation
This antibody detects mCherry.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.