Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Mouse anti-Human, DyLight 550, Clone: 857, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243769R

Catalog No. NBP243769R


Add to cart

Description

Description

mCherry Monoclonal antibody specifically detects mCherry in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

mCherry
857
Western Blot, Immunohistochemistry
Purified
Mouse
IgG1
0.1 ml
Primary
Human
Western Blot, Immunohistochemistry
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit