Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ MCHR1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer: Abnova™ H00002847T01
Specifications
Western Blot | |
MCHR1 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 46kDa | |
Transient overexpression cell lysate was tested with anti-MCHR1 antibody (Mfr. No. H00002847-D01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
293T | |
BC021146 | |
pCMV-MCHR1 full-length | |
100 μL | |
Human |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title