Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCL1 Mouse anti-Human, Clone: 8C6 + 8C6D4B1, Abnova™

Mouse monoclonal antibody raised against human MCL1.

Manufacturer:  Abnova Corporation MAB12518

Catalog No. 89-934-532


Add to cart

Description

Description

The protein encoded by this gene belongs to the Bcl-2 family. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. The longer gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene product (isoform 2) promotes apoptosis and is death-inducing. [provided by RefSeq]

Specifications

Specifications

MCL1
8C6 + 8C6D4B1
Mouse monoclonal antibody raised against human MCL1.
In PBS, pH 7.4 (0.03% sodium azide).
BCL2L3/EAT/MCL1L/MCL1S/MGC104264/MGC1839/TM
Mouse
IgG1
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Immunohistochemistry (Frozen sections) (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:1000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end
Liquid
MCL1
Recombinant protein corresponding to human MCL1.
Protein G purification
RUO
Primary
4170
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit