Learn More
MCM2 Mouse anti-Human, Clone: OTI1G4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805472
Catalog No. 50-166-7442
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).
Specifications
Specifications
MCM2
Monoclonal
Unconjugated
MCM2
Lyophilized
MCM2
Full length human recombit protein of human MCM2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1G4
PBS with 8% trehalose and no preservative; pH 7.3
P49736
BM28, CCNL1, CDCL1, D3S3194, DFNA70, MITOTIN, cdc19
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4171