Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM5 Monoclonal Antibody (4G10), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA549261

Encompass

Catalog No. PIMA549261


Add to Cart

Description

Description

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.

MCM5 Monoclonal antibody specifically detects MCM5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MCM5
Monoclonal
500 μg/mL
PBS with 4MG trehalose and no preservative
O55135, P56537, Q3KRD8
Mouse
Antigen affinity chromatography
RUO
16418, 305506, 3692
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG2a
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
4G10
Unconjugated
Eif6
AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46
E.coli-derived human EIF6 recombinant protein (Position: N66-T210).
100 μg
Primary
Human, Mouse, Rat
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit