Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Mouse anti-Human, Clone: MCM7/1466, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14845

Catalog No. 89-034-928


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human MCM7.

  • High specificity
  • High reactivity
Specifications

Specifications

MCM7
MCM7/1466
Mouse monoclonal antibody raised against partial recombinant human MCM7.
In 10 mM PBS.
Liquid
MCM7
Recombinant protein corresponding to amino acids 195-319 of human MCM7.
Protein A/G purification
RUO
Primary
4176
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined b
P33993
CDABP0042|CDC47|MCM2|P1.1-MCM3|P1CDC47|P85MCM|PNAS-146
Mouse
IgG2b
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)