Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCM8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541325
Description
Peptide sequence: IRLTEARARL ELREEATKED AEDIVEIMKY SMLGTYSDEF GNLDFERSQH Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein contains the central domain that is conserved among the MCM proteins. This protein has been shown to co-immunoprecipitate with MCM4, 6 and 7, which suggests that it may interact with other MCM proteins and play a role in DNA replication. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
MCM8 | |
Polyclonal | |
Unconjugated | |
MCM8 | |
5730432L01Rik; C20orf154; dJ967N21.5; DNA helicase MCM8; DNA replication licensing factor MCM8; MCM8; MCM8 minichromosome maintenance deficient 8; MGC119522; MGC119523; MGC12866; MGC4816; minichromosome maintenance 8; minichromosome maintenance 8 homologous recombination repair factor; minichromosome maintenance complex component 8; minichromosome maintenance deficient 8; minichromosome maintenance deficient 8 (S. cerevisiae); POF10; REC; REC homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
84515 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UJA3 | |
MCM8 | |
Synthetic peptide directed towards the C-terminal of human MCM8 (aa 717-766). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction