Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | MCM8 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1582920
|
Novus Biologicals
NBP15812920UL |
20 μL |
Each for $152.22
|
|
NBP158129
|
Novus Biologicals
NBP158129 |
100 μL |
Each for $436.00
|
|
Description
MCM8 Polyclonal specifically detects MCM8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MCM8 | |
Polyclonal | |
Rabbit | |
DNA Repair | |
C20orf154, chromosome 20 open reading frame 154, dJ967N21.5, DNA replication licensing factor MCM8, MCM8 minichromosome maintenance deficient 8, MGC119522, MGC119523, MGC12866, MGC4816, Minichromosome maintenance 8, minichromosome maintenance complex component 8, REC, REC homolog | |
MCM8 | |
IgG | |
Affinity Purified | |
94 kDa |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Q9UJA3 | |
84515 | |
Synthetic peptides corresponding to MCM8(minichromosome maintenance complex component 8) The peptide sequence was selected from the C terminal of human MCM8. Peptide sequence IRLTEARARLELREEATKEDAEDIVEIMKYSMLGTYSDEFGNLDFERSQH. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title