Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549293

Catalog No. PA549293


Add to cart

Description

Description

Predicted to react with bovine based on sequence homology.

Component of the MCM8-MCM9 complex, a complex involved in homologous recombination repair following DNA interstrand cross-links and plays a key role during gametogenesis. The MCM8- MCM9 complex probably acts as a hexameric helicase downstream of the Fanconi anemia proteins BRCA2 and RAD51 and is required to process aberrant forks into homologous recombination substrates and to orchestrate homologous recombination with resection, fork stabilization and fork restart.
Specifications

Specifications

MCM9
Polyclonal
PBS with 0.09% sodium azide
Q9NXL9
DNA helicase MCM9, hMCM9, Mini-chromosome maintece deficient domain-containing protein 1, Minichromosome maintece 9, MCM9, C6orf61, MCMDC1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
254394
Western Blot
Unconjugated
MCM9
Liquid
MCM9
KLH conjugated synthetic peptide between 316-345 amino acids from the C-terminal region of human MCM9.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit