Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCOPPB trihydrochloride, Tocris Bioscience™

Catalog Number 37631
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
1108147-88-1
C26H43Cl3N4
518.008
DTIPEVOPCGEULQ-RFCADEKQSA-N
mcoppb trihydrochloride, 1-1-1-methylcyclooctyl-4-piperidinyl-2-3r-3-piperidinyl-1h-benzimidazole trihydrochloride, mcoppb hydrochloride, mcoppb trihydrochloride/, 1108147-88-1, r-1-1-1-methylcyclooctyl piperidin-4-yl-2-piperidin-3-yl-1h-benzo d imidazole trihydrochloride, 1-1-1-methylcyclooctyl piperidin-4-yl-2-3r-piperidin-3-yl-1,3-benzodiazole trihydrochloride
25208093
1-[1-(1-methylcyclooctyl)piperidin-4-yl]-2-[(3R)-piperidin-3-yl]benzimidazole;trihydrochloride
CC1(CCCCCCC1)N2CCC(CC2)N3C4=CC=CC=C4N=C3C5CCCNC5.Cl.Cl.Cl
This item is not returnable. View return policy
1108147-88-1
C26H43Cl3N4
518.008
DTIPEVOPCGEULQ-RFCADEKQSA-N
mcoppb trihydrochloride, 1-1-1-methylcyclooctyl-4-piperidinyl-2-3r-3-piperidinyl-1h-benzimidazole trihydrochloride, mcoppb hydrochloride, mcoppb trihydrochloride/, 1108147-88-1, r-1-1-1-methylcyclooctyl piperidin-4-yl-2-piperidin-3-yl-1h-benzo d imidazole trihydrochloride, 1-1-1-methylcyclooctyl piperidin-4-yl-2-3r-piperidin-3-yl-1,3-benzodiazole trihydrochloride
25208093
1-[1-(1-methylcyclooctyl)piperidin-4-yl]-2-[(3R)-piperidin-3-yl]benzimidazole;trihydrochloride
CC1(CCCCCCC1)N2CCC(CC2)N3C4=CC=CC=C4N=C3C5CCCNC5.Cl.Cl.Cl

Potent NOP receptor agonist

High affinity NOP receptor agonist (pKi = 10.07 for the human NOP receptor). Exhibits anxiolytic effects with no effect on memory or locomotion. Regarded as one of the most potent, non-peptide NOP full agonists in vitro.
Quantity 1 mg
Formula Weight 534.05
Percent Purity >99%
Chemical Name or Material MCOPPB trihydrochloride
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit