Learn More
Invitrogen™ MCP-3 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA546952
Description
In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) MCP-1 is observed, approximately 15% cross-reactivity with rhEotaxin is observed, and less than 3% cross-reactivity with recombinant mouse Eotaxin, rhMARC, and rhMCP-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
This is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
Specifications
MCP-3 | |
Polyclonal | |
Unconjugated | |
CCL7 | |
C-C motif chemokine 7; C-C motif chemokine ligand 7; CCL7; chemokine (C-C motif) ligand 7; chemotactic protein-3; Fic; H-MCP-3; Intercrine/chemokine MARC; marc; Mcp3; MCP-3; MGC138463; MGC138465; Monocyte chemoattractant protein 3; monocyte chemotactic protein 3; NC28; Protein FIC; RANTES/sis homolog; RP23-350G1.4; SCYA6; Scya7; small inducible cytokine A7; small inducible cytokine A7 (monocyte chemotactic protein 3); small-inducible cytokine A7 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
20306 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q03366 | |
CCL7 | |
E. coli-derived recombinant mouse CCL7/MCP-3/MARC Gln24-Pro97. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.