Learn More
MCP-4 Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
$211.00 - $1836.00
Specifications
Antigen | MCP-4 |
---|---|
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P04G100
![]() ![]() |
Pepro Tech Inc
500P04G100UG |
100 μg |
Each for $270.00
|
|
|||||
500P04G1MG
![]() ![]() |
Pepro Tech Inc
500P04G1MG |
1 mg |
Each for $1,836.00
|
|
|||||
500P04G50UG
![]() ![]() |
Pepro Tech Inc
500P04G50UG |
50 μg |
Each for $211.00
|
|
|||||
Description
AA Sequence of recombinant protein: QPDALNVPST CCFTFSSKKI SLQRLKSYVI TTSRCPQKAV IFRTKLGKEI CADPKEKWVQ NYMKHLGRKA HTLKT. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human MCP-4 (CCL13). Anti-Human MCP-4 (CCL13)-specific antibody was purified by affinity chromatography employing an immobilized Human MCP-4 matrix. Sandwich ELISA: To detect Human MCP-4 (CCL13) by sandwich ELISA (using 100 μL/well) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MCP-4 (CCL13) (500-P04GBT) as a detection antibody, allows the detection of at least 2000- 4000 pg/mL of Recombinant Human MCP-4 (CCL13). Western Blot: To detect Human MCP-4 (CCL13) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MCP-4 (CCL13) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
MCP-4 Polyclonal antibody specifically detects MCP-4 in Human samples. It is validated for ELISA, Western BlotSpecifications
MCP-4 | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; C-C motif chemokine ligand 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CKb10; CK-beta-10; H-MCP-4; MCP4; MCP-4; Monocyte chemoattractant protein 4; monocyte chemotactic protein 4; NCC1; NCC-1; new CC chemokine 1; SCYA13; SCYL1; small inducible cytokine subfamily A (Cys-Cys), member 13; small-inducible cytokine A13 | |
E.coli-derived Recombinant Human MCP-4 (CCL13) | |
Antigen affinity chromatography | |
CCL13 |
ELISA, Western Blot | |
Unconjugated | |
Goat | |
Human | |
Q99616 | |
6357 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.