Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCPIP1/ZC3H12A Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30327320UL

 View more versions of this product

Catalog No. NB095416


Add to cart

Description

Description

MCPIP1/ZC3H12A Polyclonal antibody specifically detects MCPIP1/ZC3H12A in Mouse samples. It is validated for Western Blot
Specifications

Specifications

MCPIP1/ZC3H12A
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
PBS with 50% glycerol, pH7.3.
zinc finger CCCH-type containing 12A
Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human MCPIP1/ZC3H12A (NP_079355.2).
Affinity purified
RUO
Primary
80149
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit