Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCPyV T-antigen Mouse anti-Virus, Unconjugated, Clone: 2t2, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen MCPyV T-antigen
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23161MI
View Documents
MilliporeSigma
MABF2316100UL
100 μL
Each for $386.00
Add to cart
 
MABF23162MI
View Documents
MilliporeSigma
MABF231625UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

MCPyV T-antigen Monoclonal antibody specifically detects MCPyV T-antigen in Virus samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

MCPyV T-antigen
Unconjugated
Mouse
Merkel cell polyomavirus T antigen;MCV Large T antigen
Monoclonal
Protein G Purified
Immunocytochemistry, Immunohistochemistry, Western Blot
Purified
Virus
A linear peptide corresponding to 17 amino acids from the J-domain region present in both MCV LT and sT proteins.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit