Learn More
MCR Safety Acid-Resistant PVC and Nylon Coveralls
Green 0.32mm double-coated PVC on nylon scrim acid suit with attached hood
Specifications
Color Green
Seam Type Welded
Thickness 13.77 mil
View More Specs
Alert:
Caution!Do not use for fire protection. Avoid exposing PVC coveralls to flames or intense heat.
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-056-088
Accessories View Documents
MCR Safety
2981X4
4X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-056082
Accessories View Documents
MCR Safety
2981S
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-056085
Accessories View Documents
MCR Safety
2981XL
X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Provides resistance to a wide variety of chemicals, solvents and caustics. Garment is fire retardant, and cold resistant to -10°C (+14°F).
  • Midline opening is doubly protected with zipper and four-snap closure
  • Raglan sleeves, elastic-back waist
  • Graduated inseam
Specifications
Specifications
Green
Welded
13.77 mil