Learn More
MCR Safety™ Acid-Resistant PVC and Nylon Coveralls
Green 0.32mm double-coated PVC on nylon scrim acid suit with attached hood
Manufacturer:  MCR SAFETY COMPANY 2981L
 View more versions of this product
Catalog No. 19-056084
  Check Availability
Add to cart
Alert:
Caution!Do not use for fire protection. Avoid exposing PVC coveralls to flames or intense heat.
Specifications Description & Specifications
Specifications
PVC, Polyester
Welded
Large
93.98cm
37 in.
30 in.
13.77mil
1.67kg
Zipper front with double storm flap, attached hood
Green
147.32cm
33 in.
83.82cm
132.08cm
3.7 lb.
Accessories