Learn More
MCR Safety™ Acid-Resistant PVC and Nylon Coveralls
Green 0.32mm double-coated PVC on nylon scrim acid suit with attached hood
Manufacturer:  MCR SAFETY COMPANY 2981X3
 View more versions of this product
Catalog No. 19-056-087
  Check Availability
Add to cart
Alert:
Caution!Do not use for fire protection. Avoid exposing PVC coveralls to flames or intense heat.
Specifications Description & Specifications
Specifications
Zipper, Double storm flap
162.56cm
Welded
3X-Large
36 in.
91.44cm
162.56cm
4.3 lb.
PVC, Polyester
Hooded
Green
109.22cm
43 in.
32 in.
13.77mil
1.95kg
Accessories