Learn More
MCR Safety Checkmate™ Safety Glasses
Manufacturer:  MCR Safety CK110AF
 View more versions of this product
Catalog No. 18-120-488
Add to cart
Description
Description
  • Polycarbonate lens provides near total orbital seal
  • Flexible bayonet temples
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Specifications
Specifications
ANSI Compliant
Bayonet
Scratch-resistant, Anti-fog
Polycarbonate
Clear