Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Company Pigskin Driver Gloves
Pigskin gloves have excellent abrasion resistance. Offers greatest breathability because of the porous nature of the hide and becomes softer with use. Inherently retains natural softness after exposure to water.
Manufacturer: MCR Safety 3410M
Description
- The 3400 features; industry grade pigskin, straight thumb, and shirred elastic back.
- The 3410 features; premium grade pigskin, straight thumb, and shirred elastic back.
Specifications
Beige | |
carpentry, construction, driving, equipment operation, farming, horseback riding, landscaping, tractor trailer operators, wet conditions, fork lift operations | |
Brown | |
28.2 cm | |
Medium | |
13.3 cm |
28.2 x 13.3 cm (11.13 x 5.25 in.) | |
Slip-on-open | |
11.13 in. | |
Leather | |
5.25 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title