Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cow Leather Gloves with Rubberized Cuffs

Manufacturer:  MCR Safety 1310S

 View more versions of this product

Catalog No. 19-058834

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • B Grade Select Shoulder Leather
  • Blue Fabric with Red/Black Stripes
  • 4.5 in. Rubberized Safety Cuff
Specifications

Specifications

Safety
Gauntlet
Small
25.24 cm
9.94 in.
25.24 x 25.24 cm (9.94 x 9.94 in.)
Leather
9.94 in.
Black, Blue, Red
25.24 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit