Learn More
MCR Safety Crews™ Storm™ Safety Glasses
Manufacturer:  MCR Safety ST110AF
 View more versions of this product
Catalog No. 19-808-753
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Available in a variety of frame colors and lens combinations
  • Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection
  • Single wraparound dual focal point lens with integral side shields
  • 5 position ratchet action bayonet temples
  • Exclusive Duramass™ scratch resistant coating
  • Non-slip, soft temple material
Compliance
Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Specifications
Specifications
Clear
Safety
Black
Polycarbonate
Anti-fog, Scratch-resistant
Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Nylon