MCR Safety™ Crews™ Storm™ Safety Glasses
Manufacturer: MCR SAFETY COMPANY ST128
 View all options for this product
Catalog No. 19-003-049
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Certifications/Compliance Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
    Lens Material Polycarbonate
    Lens Tint Blue Mirror
    Frame Color Navy Blue
    Lens Coating Scratch-resistant
    Adjustable, nonslip temples
    This lightweight design offers extreme comfort in face-flattering eyewear. Ratchet-action temples with nonslip rubber head grips provide a secure, customized fit. The wraparound lens is constructed of scratch-resistant polycarbonate with a choice of clear or gray tint. Available in three frame colors.
  • Description & Specifications
Related Products
Product Content Feedback