Learn More
MCR Safety Cyclone PVC/Nylon Rainwear
Manufacturer:  MCR Safety 400PWXL
 View more versions of this product
Catalog No. 19-818-215
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 400BP is a bib style pant option with snap fly front, take up snaps on each ankle, and partial elastic suspenders
  • 400PW is an elastic waist pant option with snap fly front and take up snaps on each ankle
  • 400J is a jacket with detachable drawstring hood, snap front, take up snaps on the wrists
  • Nylon material is an excellent choice for tough and rugged garments
  • .35mm PVC/Nylon/PVC, rip-resistant wavelock nylon pattern
  • Snap fly front
  • Take up snaps on each ankle and on waist
  • No Pockets
  • Limited flammability, NOT intended for use as protection against ARC Flash or Flash
  • Fire
Specifications
Specifications
Yellow
X-Large
81.3cm
50 in.
Nylon, PVC
32 in.
400 Waist Pants