Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Select Grain Unlined Cowhide Leather Drivers Gloves

Gloves are constructed from unlined select grain cowhide leather with a shirred elastic back, cotton hem, and keystone thumb pattern.

$47.50 - $198.00

Specifications

ANSI Abrasion Rating ANSI 3
Back Material Shirred Elastic
Color Beige
Grade Select
Hand Specific
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-058775
View Documents
MCR Safety
3211S
Small
Dozen of 12 for $189.00
Case of 10 DZ for $1,796.00
Add to cart
 
19-058774
View Documents
MCR Safety
3211M
Medium
Dozen of 12 for $47.50
Case of 10 DZ for $1,002.50
Add to cart
 
19-058773
View Documents
MCR Safety
3211L
Large
Dozen of 12 for $189.00
Case of 10 DZ for $1,796.00
Add to cart
 
19-050-251
View Documents
MCR Safety
3211XL
X-Large
Dozen of 12 for $198.00
Case of 10 DZ for $1,877.00
Add to cart
 
Description

Description

  • Select Grade Grain Cowhide
  • Keystone Thumb
  • Shirred Elastic Back
  • Color-coded hem to identify size
Specifications

Specifications

ANSI 3
Beige
Specific
Driver Gloves
Carpentry, Farming, Landscaping, Transportation, Agriculture, and Construction
Brown
Polybag
ANSI/ISEA 105-16 Level 3
Keystone
Shirred Elastic
Select
Grain Cowhide Leather
Gunn
Open
Unlined
Leather
3211
SDS
Product Certifications

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit