Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety™ Grain Cow Drivers Gloves

For use in any general purpose application requiring greater dexterity

Manufacturer:  MCR SAFETY COMPANY 3211XL

 View more versions of this product

Catalog No. 19-050-251



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

8.17 x 6.37 in. (20.7 x 16.17cm)
Brown
Leather
6.37 in.
Beige
20.7cm
Slip-on
carpentry, construction, driving, equipment operation, farming, horseback riding, landscaping, tractor trailer operators, wet conditions, fork lift operations
X-Large
16.17cm
8.17 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit