MCR Safety™ Kevlar™ Gloves with PVC Dots
Manufacturer: MCR SAFETY COMPANY 9365S
 View more versions of this product
Catalog No. 18-999-2278
  Check Availability
Add to Cart
    Specifications
    Material Kevlar™, PVC
    Glove Cuff Style Knit
    Hem Color White
    For Use With (Application) air conditioning manufacturing, assembly line, cut protection, flame protection, food service, Glass handling, HVAC, Knife handling, light heat protection, warehousing
    Length (English) 9 in.
    Length (Metric) 23cm
    Size Small
    Coating Material PVC dots two sides
    Color Yellow
    Cut Resistance Rating Level 3
  • Description & Specifications
Related Products
Product Content Feedback