Learn More
MCR Safety Luminator PVC/Polyester Rainwear
Manufacturer:  MCR Safety 2013XL
 View more versions of this product
Catalog No. 19-181-195
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Classic Series Material is the most popular in the Industry. It combines the strength and waterproof qualities of PVC with the durability of a polyester backing. For applications where being seen is of the utmost importance, MCR Safety's Luminator™ Series offers you a variety of high visibility garments made in different colors and different options of reflective material.
  • .35 mm PVC on polyester suit, fluorescent orange
  • Lime/Silver reflectve stripes
  • Jacket with detachable hood and bib style pants
  • 2 patch pockets on jacket
  • Take up snaps on the wrist, ankles, and waist
Specifications
Specifications
Fluorescent Orange
X-Large
36.8cm
2.0
57 in.
26.6cm
Polyester/PVC
14.5 in.
10.5 in.
144.7cm
50 in.