Learn More
MCR Safety Memphis™ Leather Drivers Gloves
Manufacturer:  MCR Safety 3220S
 View more versions of this product
Catalog No. 19-058-696
Add to cart
Description
Description
The 3611 series features premium grade grain goatskin leather, shirred elastic back, and keystone thumb.
  • Premium Grade Grain Goatskin
  • Keystone Thumb
  • Shirred Elastic Back
  • Color-coded hem to identify size
Specifications
Specifications
Safety
Slip-on-open
Leather
Straight
14.75 cm
5.66 in.
14.3 x 14.75 cm (5.66 x 5.81 in.)
carpentry, construction, driving, equipment operation, farming, horseback riding, landscaping, tractor trailer operators, wet conditions, fork lift operations
Small
5.81 in.
Beige
14.3 cm