Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Memphis™ Glove™ Cotton Canvas Gloves

$12.75 - $13.50

Specifications

Color Natural
Material Canvas
Size Large
For Use With (Application) carpentry, construction, general purpose, landscaping, light heat protection, Oil and Gas industry
Glove Cuff Style Knit
View More Specs

Products 2
Catalog Number Mfr. No. Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (English) Weight (Metric) Price Quantity & Availability  
19-049056
View Documents
MCR Safety
8200CD
10 oz. 283.4 g
Dozen of 12 for $13.50
Case of 25 DZ for $337.50
Add to cart
 
19-049059
View Documents
MCR Safety
8300C
12 oz. 340.1 g
Dozen of 12 for $12.75
Add to cart
 
Description

Description

  • 100% Cotton
  • 10 ounce corded canvas material
  • Clute pattern with wing thumb
  • Knit Wrist
Specifications

Specifications

Natural
Large
Knit
Canvas
carpentry, construction, general purpose, landscaping, light heat protection, Oil and Gas industry
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit