Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Predator™ Over the Knuckle Nitrile Coated Work Gloves

Catalog Number 19041919
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Nitrile-coated gloves feature a cotton jersey lining for comfort, and a safety cuff for increased protection. Acti-Fresh™ treatment helps prevent odors while also eliminating bacteria.

  • Premium nitrile coating on the palm, fingers, and over knuckles
  • Soft jersey lining for increase comfort
  • Safety cuff for additional wrist protection
  • Treated with ActiFresh™ to eliminate odors and bacteria
  • Materials comply with all relevant federal regulations for food contact
Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Grade Premium
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Agriculture, Carpentry, Construction, Demolition, Forestry, Landscaping, and Material Handling
Glove Cuff Color Natural
Glove Cuff Style Safety
Packaging Type Polybag
Series 9760
Thumb Style Straight
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit