Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Mustang Premium Grain Leather Palm Work Gloves - Safety Cuff
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Premium Grain Cowhide Leather Palm
- 2.5 Inch Rubberized Safety Cuff
- Wing Thumb
Specifications
ANSI Abrasion Rating | ANSI 3 |
Cuff Material | Rubberized |
Color | Natural |
Grade | Premium |
Hand | Specific |
Material | Grain Cowhide Leather |
Product Type | General Purpose Gloves |
Size | Medium |
Style | Gunn |
Glove Cuff Color | Blue |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction