Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Ninja™ Flex Latex Coated Nylon Work Gloves
Latex coating on the palm and fingers enhances grip capabilities, while Actifresh™ treatment prevents odors and bacteria.
$63.10
Specifications
ANSI Abrasion Rating | ANSI 3 |
---|---|
Coating Material | Latex |
Color | Red, Gray |
Glove Cuff Style | Knit |
Material | Nylon |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-813-621
|
MCR Safety
N9680S |
Small |
Dozen of 12 for $63.10
|
|
19-813-620
|
MCR Safety
N9680M |
Medium |
Dozen of 12 for $63.10
|
|
19-813-650
|
MCR Safety
N9680L |
Large |
Dozen of 12 for $63.10
|
|
19-813-622
|
MCR Safety
N9680XL |
X-Large |
Dozen of 12 for $63.10
|
|
Description
- 15 gauge nylon shell is comfortable and flexible
- Gray latex coating on the palm and fingertips enhances grip capabilities
- Component materials comply with all relevant federal regulations for food contact
Specifications
ANSI 3 | |
Red, Gray | |
Nylon | |
15 |
Latex | |
Knit | |
Ninja™ N9680 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title