Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Ninja™ X Bi-Polymer Coated Nylon Lycra™ Work Gloves

Catalog Number p-3659546
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Bi-polymer coated gloves provide an assortment of dexterity, sense-of-touch, and protection. Actifresh™ treatment helps to prevent odors and bacteria.

  • 15 gauge Nylon Lycra™ shell increases dexterity
  • Bi-Polymer coating provides abrasion resistance
  • Actifresh™ treatments prevents odors and eliminates bacteria
  • Dark color hides dirt and grime
ANSI Abrasion Rating ANSI 4
Coating Material Bi-Polymer
Color Black
Hand Specific
Material Nylon, Lycra
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Automotive Assembly, Landscaping, Material Handling, and Warehousing
Glove Cuff Style Knit
Series Ninja™ N9674
Yarn Gauge 15
Length (English) 12.6 in.
Length (Metric) 32 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit