MCR Safety™ Nitrile Impregnated Gloves

Manufacturer: MCR SAFETY COMPANY 9710L

 View more versions of this product

Catalog No. 19-058740

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) abrasive applications, assembly, carpentry, gardening, general purpose, manufacturing, multi-purpose
    Liner Material Cotton Jersey
    Exterior Finish Smooth
    Glove Cuff Style Slip-on
    Hem Color Gold
    Material Nitrile
    Size Large
    Color Beige, blue, white

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit