Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety™ Nitrile Rubber Gloves

Manufacturer:  MCR SAFETY COMPANY 9780L

 View more versions of this product

Catalog No. 19-050-605



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Textured
Knit
Interlock Cotton Jersey
Large
15cm
CE, FDA
11 in.
11 x 5.93 in. (27.9 x 15cm)
abrasive applications, assembly, carpentry, gardening, general purpose, manufacturing, multi-purpose
Nitrile
5.93 in.
19mil
Blue,White
27.9cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit