Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety PVC Double Dip Gloves

Manufacturer:  MCR Safety 6212SJ

 View more versions of this product

Catalog No. 19-813-563


Add to cart

Description

Description

The 6212 is an industry standard black single dipped, smooth finish, interlock lined glove that is 12 inches in length.

The 6414 features a premium double dipped sandy finish PVC, jersey lining, nitrile reinforced, and 14 inch length.

  • Premium PVC
  • Green Double Dipped
  • Jersey Lined
  • Sandy finish
  • 14 Inch
Specifications

Specifications

5.6 x 6.65 in. (14.2 x 16.9cm)
Gauntlet
Jersey
Large
16.9cm
Black
14.2cm
Sandy
agriculture, battery maintenance, chemical processing, fisheries, general maintenance, mild chemical protection, Oil and Gas industry
PVC (Polyvinyl Chloride)
6.65 in.
22mil
5.6 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit