Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Red Ram™ Jersey-Lined Premium Side-Split Leather Welding Gloves
Click to view available options
X-Large
Description
- Premium Side Split Leather
- Sewn with Aramid Fiber
- One Piece, 13 in. Back
- Fully Lined and Welted
Specifications
Coating Material | Uncoated |
Color | Red |
Grade | Premium |
Material | Side Split Cowhide Leather |
Size | X-Large |
Style | Gunn |
For Use With (Application) | Construction, Foundry, Heliarc, Mig/Tig, Ship Building, Steel Work, and Welding |
Glove Cuff Style | Gauntlet |
Liner Material | Cotton Jersey |
Series | 4720 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction