MCR Safety™ String Knit Gloves

Manufacturer: MCR SAFETY COMPANY 9642SM

 View more versions of this product

Catalog No. 19-813-578

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    Hem Color Yellow
    For Use With (Application) refrigerated areas, food processing, general maintenance, parts handling, shipping/receiving and automotive plants
    Length (English) 9 in.
    Length (Metric) 23cm
    Material Cotton
    Size Small
    Color Multi color

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit