MCR Safety™ UltraTech™ Dyneema™ Cut-Resistant Gloves
Allow manual dexterity
Manufacturer: MCR SAFETY COMPANY 9676S
 View more versions of this product
Catalog No. 19-812-603
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Knit
    Hem Color Yellow
    For Use With (Application) abrasive applications, assembly, brick handling, construction, farming, general maintenance, landscaping, plumbing, recycling, shipping/receiving
    Length (English) 1 in.
    Length (Metric) 2.54cm
    Material Dyneema™
    Size Small
    Coating Material Rough
    Color Gray
    Cut Resistance Rating Level 2
  • Description & Specifications
Related Products
Product Content Feedback