Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Stretch Vinyl Impregnated Women's Gloves

Catalog Number 19082992
Click to view available options
Small
This item is not returnable. View return policy
This item is not returnable. View return policy

Impregnated vinyl gloves with a comfortable, slip-on design provide workers with basic protection.

  • Cut and Sewn construction
  • Impregnated Stretch Vinyl material
  • Single dipped
  • Slip-on cuff
  • Clute pattern
Construction Cut & Sewn
Coating Material Vinyl
Color Brown
Grade Industrial
Hand Specific
Product Type Seamless Knit Gloves
Size Small
Style Clute
For Use With (Application) Automotive Assembly, and Carpentry
Gender Women's
Glove Cuff Style Open
Series 9810
Thumb Style Straight
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit