MCR Safety™ Vinyl Impregnated Gloves

Manufacturer: MCR SAFETY COMPANY 9850M

 View more versions of this product

Catalog No. 19-049585

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Non-stretch
    Glove Cuff Style Slip-on
    Hem Color Green
    For Use With (Application) assembly, carpentry, gardening, general purpose, manufacturing, multi-purpose
    Material Vinyl
    Liner Material Cotton
    Size Medium
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit