Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCRS1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548803

Catalog No. PA548803

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity.
Specifications

Specifications

MCRS1
Polyclonal
PBS with 0.09% sodium azide
Q96EZ8
Microspherule protein 1, 58 kDa microspherule protein, Cell cycle-regulated factor p78, INO80 complex subunit J, MCRS2, MCRS1, INO80Q, MSP58
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10445
Western Blot
Unconjugated
MCRS1
Liquid
MCRS1
KLH conjugated synthetic peptide between 35-63 amino acids from the N-terminal region of human MCRS1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit