Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen MCT2
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092465
View Documents
Novus Biologicals
NBP302988100UL
100 μg
Each for $320.71
Add to cart
 
NB092466
View Documents
Novus Biologicals
NBP30298820UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

MCT2 Polyclonal antibody specifically detects MCT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

MCT2
Unconjugated
Rabbit
Lipid and Metabolism
PBS with 50% glycerol, pH7.3.
9194
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse, Rat
MCT 2, MCT2solute carrier family 16 (monocarboxylic acid transporters), member 7, monocarboxylate transporter 2, Solute carrier family 16 member 7, solute carrier family 16, member 7 (monocarboxylic acid transporter 2)
A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human MCT2 (NP_001257551.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit