Learn More
MCTP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 175781AP
Catalog No. 50-173-1661
Add to cart
Description
Description
The function of this protein remains unknown.
Specifications
Specifications
MCTP2
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5RJH2, Q6DN12
MCTP2
Rabbit
IgG
150 μL
-20°C
Primary
244049, 308742, 55784
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MCTP2
Liquid
MCTP2
MCTP2 Fusion Protein Ag11089
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat