Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCTS1 Mouse anti-Human, Clone: OTI 2G2, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502418S
Description
MCTS1 play a role in cell cycle regulation; decreases cell doubling time and anchorage-dependent growth; shortens the duration of G1 transit time and G1/S transition.Specifications
MCTS1 | |
Monoclonal | |
0.87 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9ULC4 | |
MCT-1, MCT1 | |
Mouse | |
IgG2a | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
28985 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI 2G2 | |
Unconjugated | |
MCTS1 | |
Liquid | |
MCTS1 | |
Human recombit protein fragment corresponding to amino acids 1-181 of human MCTS1 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title