Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCTS1 Mouse anti-Human, Clone: OTI 2G2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502418

Catalog No. 50-167-3479


Add to cart

Description

Description

MCTS1 play a role in cell cycle regulation; decreases cell doubling time and anchorage-dependent growth; shortens the duration of G1 transit time and G1/S transition.
Specifications

Specifications

MCTS1
Monoclonal
0.87 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9ULC4
MCT-1, MCT1
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
28985
Immunohistochemistry (Paraffin), Western Blot
OTI 2G2
Unconjugated
MCTS1
Liquid
MCTS1
Human recombit protein fragment corresponding to amino acids 1-181 of human MCTS1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit