Learn More
MCUR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 249481AP
Catalog No. 50-173-4637
Add to cart
Description
Description
Mcur1 is a key regulator of mitochondrial calcium uniporter (MCU) required for calcium entry into mitochondrion. Mcur1 plays a direct role in uniporter-mediated calcium uptake via a direct interaction with MCU (mitochondrial calcium uniporter). Mcur1 may be involved in the assembly of the membrane components of the uniporter complex.
Specifications
Specifications
MCUR1
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96AQ8, Q9CXD6
C6orf79, CCDC90A, MCUR1
Rabbit
IgG
150 μL
-20°C
Primary
63933, 76137
Immunoprecipitation, Western Blot
Unconjugated
MCUR1
Liquid
MCUR1
MCUR1 Fusion Protein Ag20040
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse