missing translation for 'onlineSavingsMsg'
Learn More

MDA 19, Tocris™

CB2 agonist

Manufacturer:  R&D Systems 4205/50

Quantity 50mg

 View more versions of this product

Catalog No. 42-055-0

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

>99%
349.43
MDA 19
C21H23N3O2
N'-[(3Z)-1-Hexyl-2-oxo-1, 2-dihydro-3H-indol-3-ylidene]benzohydrazide

Specifications

50mg
>99%
349.43
MDA 19
1048973-47-2
C21H23N3O2
N'-[(3Z)-1-Hexyl-2-oxo-1, 2-dihydro-3H-indol-3-ylidene]benzohydrazide

Safety and Handling

Recommended Storage : Store at +4°C

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'