Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDA5 Mouse anti-Human, Alexa Fluor™ 488, Clone: OTI8C10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272604AF488
Description
MDA5 Monoclonal specifically detects MDA5 in Human samples. It is validated for Western Blot.Specifications
MDA5 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IFIH1 | |
Full length recombinant protein of human MDA5 (NP_071451) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI8C10 | |
Western Blot | |
CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
64135.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title